missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoamine Oxidase B Antibody (8E0), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42993
This item is not returnable.
View return policy
Description
Monoamine Oxidase B Monoclonal antibody specifically detects Monoamine Oxidase B in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Monoamine Oxidase B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 8 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| adrenalin oxidase, amine oxidase [flavin-containing] B, EC 1.4.3, EC 1.4.3.4, MAO, brain, MAO, platelet, MAO-B, MGC26382, monoamine oxidase B, Monoamine oxidase type B, tyramine oxidase | |
| monoamine oxidase B - (Uniprot# P27338) | |
| 100 μg | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4129 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction