missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals

Product Code. 10073240
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38330-80-2
Molecular Formula C4H5O4·K
Molecular Weight (g/mol) 156.18
MDL Number MFCD00014021
InChI Key WWTULTKUWBKVGV-UHFFFAOYSA-M
Synonym potassium 3-methoxy-3-oxopropanoate, methyl potassium malonate, potassium methyl malonate, propanedioic acid, monomethyl ester, potassium salt, potassium monomethyl malonate, monomethyl malonate potassium salt, hydrogen methyl malonate potassium salt, monomethylpotassiummalonate, acmc-209vp4, ksc226a9d
PubChem CID 2724687
IUPAC Name potassium;3-methoxy-3-oxopropanoate
SMILES COC(=O)CC(=O)[O-].[K+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >200°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula COOKCH2COOCH3
Quantity 25 g
Fieser 15,207
Formula Weight 156.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl monopotassium malonate
Show More Show Less
compliance-icons
Product Identifier
  • Monomethyl monopotassium malonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'