missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera
Click to view available options
Grade:
Antigen Factor 2
Antigen Factor 5
Antigen Factor d
Antigen Factor eh
Antigen Factor enx
Antigen Factor f
Antigen Factor gm
Antigen Factor h
Antigen Factor k
Antigen Factor l, v
Antigen Factor l, w
Antigen Factor q
Antigen Factor s
Antigen Factor t
Antigen Factor u
Antigen Factor z13
Antigen Factor z24
Antigen Factor z29
Antigen Factor z32
Antigen Factor z6
Unit Size:
Each
Description
While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.
- Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
- Produced using reference strains prepared by World Health Organization guidelines
- Certified reference center performance
- Presented in color-coded dropper bottles
- Long shelf life: up to 5 years from date of manufacture
Specifications
Specifications
| Certifications/Compliance | W.H.O. |
| Content And Storage | 2°C to 8°C, Do Not Freeze |
| Grade | Antigen Factor eh |
| Product Type | Antiserum |
| For Use With (Application) | Traditional Slide Agglutination Testing |
| Packaging Type | Dropper Bottle |
| Quantity | 3.0 mL |
| Slide Type | Agglutination |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur