missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera
Click to view available options
Grade:
Antigen Factor 2
Antigen Factor 5
Antigen Factor d
Antigen Factor eh
Antigen Factor enx
Antigen Factor f
Antigen Factor gm
Antigen Factor h
Antigen Factor k
Antigen Factor l, v
Antigen Factor l, w
Antigen Factor q
Antigen Factor s
Antigen Factor t
Antigen Factor u
Antigen Factor z13
Antigen Factor z24
Antigen Factor z29
Antigen Factor z32
Antigen Factor z6
Unit Size:
Each
Description
While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.
- Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
- Produced using reference strains prepared by World Health Organization guidelines
- Certified reference center performance
- Presented in color-coded dropper bottles
- Long shelf life: up to 5 years from date of manufacture
Specifications
Specifications
| Certifications/Compliance | W.H.O. |
| Content And Storage | 2°C to 8°C, Do Not Freeze |
| Grade | Antigen Factor s |
| Product Type | Antiserum |
| For Use With (Application) | Traditional Slide Agglutination Testing |
| Packaging Type | Dropper Bottle |
| Quantity | 3.0 mL |
| Slide Type | Agglutination |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction