missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera

Use in slide agglutination testing for identification of Salmonella species/antigens according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera is a comprehensive range of monovalent Salmonella antisera delivered in economical 3.0mL bottles.

Brand:  Pro-Lab Diagnostics™ PL6025

 View more versions of this product

Product Code. 11909074

  • £96.00 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.

  • Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
  • Produced using reference strains prepared by World Health Organization guidelines
  • Certified reference center performance
  • Presented in color-coded dropper bottles
  • Long shelf life: up to 5 years from date of manufacture
Specifications

Specifications

W.H.O.
Species: Group G2, Antigen Factor 23
Traditional Slide Agglutination Testing
3.0 mL
2°C to 8°C, Do Not Freeze
Antiserum
Dropper Bottle
Agglutination
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers