missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera

Use in slide agglutination testing for identification of Salmonella species/antigens according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera is a comprehensive range of monovalent Salmonella antisera delivered in economical 3.0mL bottles.

Brand:  Pro-Lab Diagnostics™ PL6018

 View more versions of this product

Product Code. 11939064

  • £220.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.

  • Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
  • Produced using reference strains prepared by World Health Organization guidelines
  • Certified reference center performance
  • Presented in color-coded dropper bottles
  • Long shelf life: up to 5 years from date of manufacture
Specifications

Specifications

W.H.O.
Species: Group E1, Antigen Factors 10
Traditional Slide Agglutination Testing
3.0 mL
2°C to 8°C, Do Not Freeze
Antiserum
Dropper Bottle
Agglutination
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.