missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS 10X Solution, Fisher BioReagents™
Premixed MOPS buffer solution (pH = 7.0) at 10X concentration. A low ionic strength buffer used for electrophoretic separation of formaldehyde denatured RNA
£22.11 - £30.07
Chemical Identifiers
CAS | 1132-61-2, 7732-18-5, 1310-73-2 |
---|---|
Molecular Formula | C7H15NO4S |
Molecular Weight (g/mol) | 209.26 |
MDL Number | MFCD00006183 |
InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
Synonym | 3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-morpholinopropanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, unii-273bp63nv3, 3-n-morpholino propanesulfonic acid, morpholinopropanesulfonic acid, 3-[N-Morpholino]propane sulfonic Acid, 4-morpholinepropanesulfonic acid |
PubChem CID | 70807 |
ChEBI | CHEBI:44115 |
IUPAC Name | 4-(3-sulfonatopropyl)morpholin-4-ium |
SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
Description
Premixed MOPS buffer solution (pH = 7.0) at 10X concentration. A low ionic strength buffer used for electrophoretic separation of formaldehyde denatured RNA
A low ionic strength buffer used for electrophoretic separation of RNAChemical Identifiers
1132-61-2, 7732-18-5, 1310-73-2 | |
209.26 | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
70807 | |
4-(3-sulfonatopropyl)morpholin-4-ium |
C7H15NO4S | |
MFCD00006183 | |
3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-morpholinopropanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, unii-273bp63nv3, 3-n-morpholino propanesulfonic acid, morpholinopropanesulfonic acid, 3-[N-Morpholino]propane sulfonic Acid, 4-morpholinepropanesulfonic acid | |
CHEBI:44115 | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
Specifications
1132-61-2, 7732-18-5, 1310-73-2 | |
DNase free | |
C7H15NO4S | |
Protease free | |
3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-morpholinopropanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, unii-273bp63nv3, 3-n-morpholino propanesulfonic acid, morpholinopropanesulfonic acid, 3-[N-Morpholino]propane sulfonic Acid, 4-morpholinepropanesulfonic acid | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
209.26 | |
CHEBI:44115 | |
MOPS, 10X Solution, pH 7 |
Liquid | |
0.2-micron filtered | |
MFCD00006183 | |
15, 6352 | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
4-(3-sulfonatopropyl)morpholin-4-ium | |
70807 | |
10X solution contains 0.2M MOPS, 20mM sodium acetate trihydrate, 10mM disodium EDTA and NaOH to pH of 7.0 |
Safety and Handling
missing translation for 'shelfLife' : 1 year
Recommended Storage : 4°C