missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shield Medicare™ Mops
£295.00 - £473.00
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12897173
|
Shield Medicare™
10056355 |
1400 mm |
£473.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
12837183
|
Shield Medicare™
3036300 |
1000 to 1500 mm |
£295.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Specifications
Mop | |
Autoclavable | |
1 Ea. |