missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99.5%, for molecular biology, Dnase, Rnase, Prot. free, for peptide sequencing

Product Code. p-3734348 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10142871

Brand: Thermo Scientific Chemicals 327661000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Absorbance 0.01 max. at 280nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 277.0°C to 282.0°C
CAS Min % 99.4
CAS Max % 100.0
Color White
pH 3.0 to 4.5 (1% soln. at 25°C)
Physical Form Powder
Quantity 100 g
Flash Point 116°C
Assay Percent Range 99.50%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C)
Packaging Glass bottle
Merck Index 15, 6352
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: <10g/L ethanol (20°C)
Formula Weight 209.26
Percent Purity 99.5%
Grade Molecular Biology
Chemical Name or Material MOPS, DNAse, RNAse and Protease Free
Show More Show Less
Product Identifier
  • MOPS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.