missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, 99%, Thermo Scientific Chemicals
18.53 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 277°C to 280°C (decomposition) |
Quantity | 25 g |
Flash Point | 110°C (230°F) |
Assay Percent Range | 99% |
Beilstein | 1106776 |
Merck Index | 14,6265 |
Solubility Information | Soluble in water. |
IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
Formula Weight | 209.27 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'