missing translation for 'onlineSavingsMsg'
Learn More

MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Commonly used buffering agent

£85.00 - £240.00

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(4-Morpholino)propane sulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10234673
SDS View Documents Promotion Details
Fisher BioReagents
BP308-100
100 g Glass Bottle
£85.00
100g
Please to purchase this item. Need a web account? Register with us today!
10234723
SDS View Documents Promotion Details
Fisher BioReagents
BP308-500
500 g Poly Bottle
£240.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Commonly used buffering agent

This biological buffer has a usable pH range of 6.5 to 7.9.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.