missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, JustPURE™, white powder, Fisher BioReagents
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Specifications
Absorbance | 0.015 max. at 280nm |
CAS | 1132-61-2 |
Melting Point | 277°C |
Color | White |
Decomposition Information | >277°C |
pH | 2.5 to 4.0 |
Physical Form | Powder or Solid |
Quantity | 100 g |
Assay Percent Range | ≥99.5 % |
Loss on Drying | 1% max. |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- MOPS
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'