missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10391284
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
MDL Number MFCD00064350
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-(4-Morpholino)propanesulfonic acid sodium salt
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Plastic bottle
Formula Weight 231.25
Percent Purity 98%
Chemical Name or Material MOPS sodium salt, 98%
Show More Show Less
Product Identifier
  • MOPS, sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.