missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS sodium salt, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.
Solubility
Soluble in water (523 g/L at 20°C).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.
Solubility
Soluble in water (523 g/L at 20°C).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
Melting Point | ∽260°C (decomposition) |
Quantity | 25 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in water (523g/L at 20°C). |
SMILES | C1COCCN1CCCS(=O)(=O)[O-].[Na+] |
Molecular Weight (g/mol) | 231.242 |
Formula Weight | 231.25 |
Chemical Name or Material | MOPS sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'