Learn More
Morpholine, 99+%, ACS reagent, Thermo Scientific Chemicals
£19.70 - £170.00
Chemical Identifiers
CAS | 110-91-8 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00005972 |
InChI Key | YNAVUWVOSKDBBP-UHFFFAOYSA-N |
Synonym | 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro |
PubChem CID | 8083 |
ChEBI | CHEBI:34856 |
IUPAC Name | morpholine |
SMILES | C1COCCN1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055710
|
Thermo Scientific Acros
415160050 |
5 g | Glass bottle |
£19.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209940
|
Thermo Scientific Acros
415165000 |
500 g | Glass Bottle |
£50.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10258853
|
Thermo Scientific Acros
415160025 |
2.5 kg | Glass bottle |
£170.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-91-8 | |
87.12 | |
YNAVUWVOSKDBBP-UHFFFAOYSA-N | |
8083 | |
morpholine |
C4H9NO | |
MFCD00005972 | |
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro | |
CHEBI:34856 | |
C1COCCN1 |
Specifications
110-91-8 | |
100.0 | |
0.9900g/mL | |
32°C | |
99+% | |
1.4530 to 1.4550 | |
27, 5 | |
15, 6362 | |
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro | |
YNAVUWVOSKDBBP-UHFFFAOYSA-N | |
morpholine | |
8083 | |
2.23 mPa.s (20°C) | |
11mbar at 20°C | |
ACS Reagent | |
Morpholine, 99+% |
99.0 | |
-5.0°C | |
126.0°C to 130.0°C | |
Authentic | |
C4H9NO | |
MFCD00005972 | |
01,705; 11,352; 13,14 | |
0.99 | |
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil | |
C1COCCN1 | |
87.12 | |
CHEBI:34856 | |
87.12 | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-815-1
missing translation for 'rtecsNumber' : QD6475000
missing translation for 'tsca' : TSCA
- Morpholine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only