missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morpholino(4-piperidinyl)methanone hydrochloride, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Spécification
Spécification
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 4-(piperidine-4-carbonyl)morpholine |
Formula Weight | 234.73 |
Percent Purity | 90% |
Chemical Name or Material | Morpholino(4-piperidinyl)methanone hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'