Learn More
Motic™ Moticam A2
Give yourself a head start into digital microscopy with steady 30 fps and a 1080p resolution. The Moticam A2 is a great choice for anyone who wants to begin their journey into live imaging and sharing their discoveries.
Brand:  Motic™ 1100600101192
Product Code. 17847960
  • £289.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Live Display Modes: 1920 x 1080 px, 1280 x 720 px
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
Specifications
Specifications
Camera
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap
1/3.1 in.
Moticam A
sCMOS
30 fps
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ Moticam A2 > Moticam A2; Frequency (Frame Rate): 30 fps; Sensor Types: sCMOS; Product Line: Moticam A Series; Includes: Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap