Learn More
Motic™ Moticam A8
The fast sCMOS imaging setup of the Moticam A8 allows 4K resolution while delivering steady 15 fps. Printing images requires high-resolution data; Moticam delivers the detail needed.
Brand:  Motic™ 1100600101443
Product Code. 17877960
  • £436.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The ideal camera, for everyone who has high demands and wants to stay flexible.
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
  • Live Display Modes: 3840 x 2160 px, 1920 x 1080 px
  • Maximum Frame Rate: 3840 x 2160 at 15 fps, 1920 x 1080 at 30 fps
Specifications
Specifications
Camera
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap
1/3 in.
Moticam A
sCMOS
15 to 30 fps
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ Moticam A8 > Moticam A8; Frequency (Frame Rate): 15 to 30 fps; Sensor Types: sCMOS; Product Line: Moticam A Series; Includes: Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap