missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Moulded Polyethylene Connecting Hook

Used to attach lengths of chain, our durable B-900 Moulded Polyethylene Connecting Hooks are available to match our posts and chain.

Brand:  Brady™ 078257

 View more versions of this product

Product Code. 11781296

  • £52.95 / 1 pack
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used to attach lengths of chain, our durable B-900 polyethylene Connecting Hooks are available to match our posts and chain.
Specifications

Specifications

36 Pk.
Yellow
Traffic Control
50.8 mm
Hook
Polyethylene
2 in.
Stanchion Accessories
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers