Learn More
SCHOTT AG Lighting and Imaging™ Mounting Bracket for EasyLED Spotlight Plus and KL300LED
Product Code. 11358330 Shop All Schott Products
Click to view available options
For Use With (Equipment):
25 mm dia. Column
30 mm dia. Column
32 mm dia. Column
35 mm dia. Column
Base 158.340
Zeiss Stand K, Stand M
Unit Size:
1 unit
Each
Product Code. 11358330
Brand: SCHOTT AG Lighting and Imaging™ 120250
Only null left
Add to basket
Add to basket
The Mounting Bracket allows ergonomic operation of Easy LED Spotlight and KL300LED. Connect directly to the microscope via column-adapter.
  • Connects the EasyLED Spotlight Plus or KL300LED to the microscope column, base or a base.
Specifications
Type Mounting Bracket
For Use With (Application) Stereo Microscopy Illumination
For Use With (Equipment) Base 158.340