missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Angiopoietin-like Protein 2 (AAD55358) VersaClone cDNA
R&D Systems™ Mouse Angiopoietin-like Protein 2 (AAD55358) VersaClone cDNA is a secreted glycoprotein.
Brand: R&D Systems™ RDC2254
Additional Details : Weight : 0.00970kg
Description
The Mouse Angiopoietin-like Protein 2 (AAD55358) VersaClone cDNA clone contains the complete ORF for Angiopoietin-like Protein 2/ANGPTL2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Angiopoietin-like Protein 2/ANGPTL2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
mAngptl2 | |
10 μg at 0.2 μg/μL | |
1495bp | |
23452 | |
Mouse |
AAD55358 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |