Learn More
Invitrogen™ Mouse ANGPTL4 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP178
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Mouse FIAF Synthetic Peptide, PEP-178, from Invitrogen.
PEP-178 is a 15 amino acid synthetic peptide whose sequence corresponds to mouse FIAF. The sequence of this peptide is (amino to carboxy terminus): C-Q-G-P-K-G-K-D-A-P-F-K-D-S-E (NH2)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and FIAF, catalog # PA1-1053. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
57875 | |
purified | |
Mouse ANGPTL4 | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
Q9Z1P8 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Angptl4 | |
No preservative | |
50 μg | |
C-Q-G-P-K-G-K-D-A-P-F-K-D-S-E(NH2) |