missing translation for 'onlineSavingsMsg'
Learn More

BARX1, Mouse, Clone: 1E7, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant BARX1.

Brand:  Abnova H00056033-M01A.200uL

Product Code. 16102033

  • £283.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene encodes a member of the Bar subclass of homeobox transcription factors. Studies of the mouse and chick homolog suggest the encoded protein may play a role in developing teeth and craniofacial mesenchyme of neural crest origin. The protein may also be associated with differentiation of stomach epithelia. [provided by RefSeq

Sequence: MGLEKRFEKQKYLSTPDRIDLAESLGLSQLQVKTWYQNRRMKWKKIVLQGGGLESPTKPKGRPKKNSIPTSEQLTEQERAKDAEKPAEVPGEPSDRSRED
Specifications

Specifications

BARX1
Monoclonal
Unconjugated
ascites with no preservative
BC009458
Mouse
200 μL
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
1E7
Mouse monoclonal antibody raised against a full-length recombinant BARX1.
BARX1
BARX1
BARX1 (AAH09458,1 a.a. ∽ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
56033
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Ascites
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.