Learn More
Mouse anti-Feline Ig Mix Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA5-10357
Additional Details : Weight : 0.01000kg
Description
SA1-10642 detects feline Immunoglobulin. Does not react with bovine, equine, canine, rabbit, sheep, porcine, donkey, guinea pig, goat or human. SA1-10642 has been successfully used in ELISA applications. The SA1-10642 immunogen is purified feline immunoglobulin.
Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Feline Ig Mix | |
Monoclonal | |
0.7 mg/mL | |
PBS with no preservative; pH 7.4 | |
Purified feline immunoglobulin. | |
100μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
ELISA | |
FIG10-118.1 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Feline | |
Liquid |