Learn More
Mouse anti-Goat IgG whole molecule, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB9210.2mL
Product Code. 16234630
  • £218.00 / 2mL
Specifications
Mouse anti-Goat IgG whole molecule
Polyclonal
Mouse anti-Goat IgG recognizes Goat IgG whole molecule. This secondary antibody was purified using delipidation and defibrination.
Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2
Goat IgG whole molecule
2 mL
Goat
Lyophilized
ELISA, Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Mouse
This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against goat IgG and goat serum.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.