missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA-tag, Mouse, Agarose, Clone: 12CA5, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of HA-tag.
Brand: Abnova MAB1912.1mL
Additional Details : Weight : 0.02000kg
Specifications
HA-tag | |
Monoclonal | |
Agarose | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mL | |
Primary | |
Liquid |
Immunoprecipitation, Western Blot | |
12CA5 | |
Mouse monoclonal antibody raised against synthetic peptide of HA-tag. | |
In 20mM sodium phosphate buffer, 150mM NaCl, pH 7.5 (3mM sodium azide) | |
A synthetic peptide corresponding to HA-tag. | |
RUO | |
Store at 4°C. Do not freeze. | |
IgG2b |