missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human IgG Fab Secondary Antibody, PE
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen™ MA110377
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody detects Fab fragments regardless light chain type.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
Human IgG Fab | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Purified human IgG. | |
100 μg | |
Secondary | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
4A11 | |
PE | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Human | |
Secondary Antibody | |
IgG2b |
Produktvorschläge
Customers who viewed this item also viewed
Viewing 1-5 of 12
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AC4667894 | Certificate of Analysis | 31/03/2025 | |
AC4664353A | Certificate of Analysis | 24/03/2025 |
Sonderangebote
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
58Hours
14Minutes
1Seconds
49New Offer Save Up to 92%
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Invitrogen™ Mouse anti-Human IgG Fab Secondary Antibody, PE > 100 μg; PE
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur