Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2046H
Product Code. 30226961
  • £261.00 / 0.20mL
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG1 Fc
Monoclonal
HRP
PBS
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum.
0.2 mL
Secondary
Store at 4°C in the dark.
IgG1
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion
2C11cc
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunodiffusion
Mouse
Protein A purified
RUO
Human
Purified