Learn More
Invitrogen™ Mouse anti-Human IgG1 Fc Secondary Antibody, Alexa Fluor™ 488
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ A10631
Product Code. 10794259
  • £240.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Previous lots were labeled as Mouse anti-Human IgG1. Both descriptions are correct. Please reference https://pubmed.ncbi.nlm.nih.gov/3069872/ for clone information. Product will be shipped at Room Temperature.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgG1 Fc
Monoclonal
1 mg/mL
PBS with 5mM sodium azide; pH 7.5
IgG gamma 1.
250 μg
Secondary
4°C, store in dark
Liquid
Flow Cytometry, Immunocytochemistry
HP6069
Alexa Fluor 488
Mouse
purified
RUO
Human
Secondary Antibody
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Mouse anti-Human IgG1 Fc Secondary Antibody, Alexa Fluor™ 488 > 250 μg; Alexa Fluor 488