Learn More
Invitrogen™ Mouse anti-Human IgG4 Fc Secondary Antibody, Biotin
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ A10663
135.75 GBP valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.
Product Code. 10365878
  • £181.00 / 250µg
Seek Out Great Savings With the Lab of Curiosity
Use promo code 24110
View all promotions
For Research Use Only. All usage must comply with product instructions.
Description
Description
Mouse anti-Human IgG4 Fc Secondary Antibody for ICC/IF, Western Blot
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgG4 Fc
Monoclonal
1 mg/mL
PBS with 5mM sodium azide; pH 7.5
Human IgG4.
250 μg
Secondary
4°C
Liquid
Western Blot, Immunocytochemistry
HP6025
Biotin
Mouse
Affinity chromatography
RUO
Human
Secondary Antibody
IgG1