missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgM Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA135473
This item is not returnable.
View return policy
Description
This antibody does not cross react with albumin fractions of human serum, human IgA, human IgG or albumin fractions from other species
Human IgM Monoclonal antibody specifically detects Human IgM in Human samples. It is validated for Dot Blot, ELISA, Immunohistochemistry (Frozen).Specifications
Human IgM | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5 | |
Purified human IgM. | |
1 mL | |
Secondary | |
-20°C | |
IgG1 |
Dot Blot, ELISA, Immunohistochemistry (Frozen) | |
ICL-931 | |
HRP | |
Mouse | |
Ion-exchange chromatography, Size-exclusion - Gel filtration | |
RUO | |
Human | |
Liquid |