Learn More
Mouse anti-Human IgM Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA514712
Code : A0
Additional Details : Weight : 0.01000kg
Description
By ELISA and WB, MA5-14712 does not react with Human IgG or Human IgA. Product MA514712 is a smaller package size of MII0402 (formerly sold as a Seradyn product).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgM | |
Monoclonal | |
Unconjugated | |
Immunoglobulin M, IgM | |
Purified Human Serum IgM | |
100 μg | |
Secondary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
IgG1 |
ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Human | |
Liquid |