missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, PE, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA110389
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with hamster, goat, guinea pig, rabbit or sheep.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human Kappa Light Chain | |
Monoclonal | |
PE | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
A8B5 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
not available | |
100 Tests | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |