Learn More
Mouse anti-Human Lambda Light Chain Secondary Antibody, PE, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA180937
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL. Mouse anti Human Lambda Light Chain antibody, clone Mc24-IC6 recognizes human lambda light chains, and shows no cross- reactivity to kappa light chains or to immunoglobulin heavy chains.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human Lambda Light Chain | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
Purified human IgG. | |
100 Tests | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |
ELISA, Flow Cytometry | |
Mc24-IC6 | |
PE | |
Mouse | |
Protein A | |
RUO | |
Human | |
Lyophilized |