Learn More
CD38 Mouse anti-Human, Unlabeled, Clone: HB7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 347680
Additional Details : Weight : 0.03000kg
Description
The CD38 antibody, clone HB7, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with the BJAB cell line.
The CD38 antibody recognizes an integral membrane glycoprotein of 45 kilodaltons (kDa), with a protein core of 35 kDa. The CD38 antigen is also known as T10, ADP-ribosyl cyclase, and cyclic ADP ribose hydrolase 1.
Specifications
CD38 | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human BJAB B cell line | |
RUO | |
952 | |
Store at 2-8°C. Protect from light. | |
IgG1 κ |
Flow Cytometry | |
HB7 | |
Unconjugated | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Purified |
For Research Use Only.