missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Mouse IgM[a], Clone: DS-1, Secondary Antibody, BD

Brand:  BD Biosciences 553514

Additional Details : Weight : 0.01000kg

Product Code. 15807258

  • £1670.00 / Each
Estimated Shipment: 30-07-2024
to see stock.

Description

Description

The DS-1 antibody reacts specifically with mouse IgM of Igh-C[a] and related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2). It does not react with IgM of Igh-C[b], Igh-C[e], or related hapltypes (e.g., C57BL, SJL, A, AKR, NZB). DS-1 an

Host Species: Mouse
Clone: DS-1
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Mouse NMRI Monoclonal Antibody TC31

ELISA, Flow Cytometry, Immunohistochemistry (Frozen)

Specifications

Specifications

Mouse anti-Mouse IgM[a]
DS-1
Mouse
0.5 mg
Immunology
Mouse
Monoclonal
0.5mg/mL
Affinity Purified
RUO
Secondary
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers