Learn More
Invitrogen™ Mouse anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody
Mouse Polyclonal Secondary Antibody
Brand: Invitrogen™ 31213
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31213 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31213 reacts with the heavy chains of rabbit IgG and with light chains of most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, goat, mouse and sheep serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Mouse | |
1.5 mg | |
Secondary | |
4°C | |
Liquid |
Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
1.8 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Secondary Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.