Learn More
Invitrogen™ Mouse anti-Rat IgE Secondary Antibody, Biotin
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ MA516811
Product Code. 15216059
  • £451.00 / 500µg
Explore more special offers
Description
Description
Mouse anti Rat IgE antibody, clone MARE-1 recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgE
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide
Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins.
500 μg
Secondary
4°C
Liquid
ELISA, Western Blot
MARE-1
Biotin
Mouse
Affinity chromatography
RUO
Rat
Secondary Antibody
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Mouse anti-Rat IgE Secondary Antibody, Biotin > 500 μg; Biotin