Learn More
Mouse anti-Rat IgM (Heavy chain) Secondary Antibody, Biotin, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA516820
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Mouse anti Rat IgM Heavy Chain antibody, clone MARM-4 recognizes the mu heavy chain of rat immunoglobulin and does not cross-react with other immunoglobulin classes.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgM (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and <0.1% sodium azide | |
IR202 rat IgM myeloma protein | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA | |
MARM-4 | |
Biotin | |
Mouse | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |