missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgM Secondary Antibody, FITC, Invitrogen™

Mouse Monoclonal Secondary Antibody

Brand:  Invitrogen SA125272

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11535050

  • £266.00 / 125µg
Estimated Shipment: 24-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SA1-25272 recognizes the mu heavy chain of rat immunoglobulin and does not cross react with other immunoglobulin classes. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Rat IgM
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide
Polyclonal rat IgM.
125 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
IgG1
Flow Cytometry
MARM-4
FITC
Mouse
Affinity chromatography
RUO
Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers