missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse APLP-2 Antibody, R&D Systems™
Goat Polyclonal Antibody
£147.94 - £418.32
Specifications
Antigen | APLP-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726827
|
R&D Systems
AF3685-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18023122
|
R&D Systems
AF3685 |
100 μg |
£418.32
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APLP-2 Polyclonal specifically detects APLP-2 in Mouse samples. It is validated for Western Blot.Specifications
APLP-2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II | |
APLP2 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAH52396.2 | |
334 | |
Mouse myeloma cell line NS0-derived recombinant mouse APLP-2, Leu37-Ser624, Accession # AAH52396.2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse APLP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) APLP-2 is observed and less than 1% cross-reactivity with rhAPLP-1 and recombinant mouse APLP-1 is observed. |
For Research Use Only