missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL2/JE/MCP-1 DuoSet ELISA
Mouse CCL2/JE/MCP-1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
£240.00 - £452.00
Specifications
Product Type | ELISA Development Kit |
---|---|
Quantity | 1 Kit |
Species | Mouse |
Conjugate | Biotin |
Assay Range | 3.91 to 250 pg/mL |
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
30000488
|
R&D Systems™
DY479 |
15 Plates |
£452.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30021773
|
R&D Systems™
DY479-05 |
5 Plates |
£240.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Mouse CCL2/JE/MCP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CCL2/JE/MCP-1.Specifications
ELISA Development Kit | |
Mouse | |
3.91 to 250 pg/mL | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
20296 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title