missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CCL22 (MDC) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £2400.00
Specifications
Accession Number | O88430 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 20299 |
Molecular Weight (g/mol) | 7.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17881813
|
Gibco™
250-23-100UG |
100 μg |
£480.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830443
|
Gibco™
250-23-1MG |
1 mg |
£2400.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17820843
|
Gibco™
250-23-20UG |
20 μg |
£157.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17841623
|
Gibco™
250-23-250UG |
250 μg |
£838.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17860723
|
Gibco™
250-23-500UG |
500 μg |
£1436.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861623
|
Gibco™
250-23-5UG |
5 μg |
£64.75
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.Specifications
O88430 | |
protein with no preservative | |
7.8 | |
E. coli | |
RUO | |
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
Mouse | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
20299 | |
Mouse CCL22 (MDC) | |
-20°C | |
<1 EU/μg | |
CCL22 | |
Unconjugated | |
GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |