missing translation for 'onlineSavingsMsg'
Learn More

GIBCO™ Mouse (CD-1) Microsomes

Product Code. 10772650
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10772650

Brand: GIBCO™ MSMCPL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Liver microsomes prepared from a pool of male CD-1 mice. Total protein concentration: 20mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Donor Source Pooled
Gender Male
Physical Form Frozen
Tissue Liver
Quantity 0.5 mL
Purity Research Grade
Host Species Mouse
Concentration 20 mg/mL
Product Type Mouse Microsome
Content And Storage Store between -60°C and -85°C.
Format Tube
Sub Cellular Localization Microsomes
Shipping Condition Dry Ice
Product Line Gibco
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.