Learn More
R&D Systems™ Mouse Cortical Stem Cells (2 x 10e6 cells/vial)
For cortical stem cell expansion and differentiation into neurons, astrocytes and oligodendrocytes
Brand: R&D Systems™ NSC002
Additional Details : Weight : 0.00970kg
Description
- Have been fully characterized to reduce experimental variation
- Have been verified to be free of mycoplasma and microbial contamination
Specifications
2 x 10e6 cells/vial |