missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CRELD2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3686-SP
This item is not returnable.
View return policy
Description
CRELD2 Polyclonal specifically detects CRELD2 in Mouse samples. It is validated for Western Blot.Specifications
CRELD2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, DKFZp667O055, MGC11256 | |
Goat | |
Affinity Purified | |
RUO | |
79174 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q9CYA0 | |
CRELD2 | |
Mouse myeloma cell line NS0-derived recombinant mouse CRELD2 Ser23-Leu350 Accession # Q9CYA0 | |
25 μg | |
Primary | |
Detects mouse CRELD2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CRELD2 is observed and less than 1% cross-reactivity with recombinant mouse CRELD1 is observed. | |
Mouse | |
IgG |
For Research Use Only