missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CRELD2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
£147.94 - £418.32
Specifications
Antigen | CRELD2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18706567
|
R&D Systems
AF3686-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18039591
|
R&D Systems
AF3686 |
100 μg |
£418.32
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRELD2 Polyclonal specifically detects CRELD2 in Mouse samples. It is validated for Western Blot.Specifications
CRELD2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, DKFZp667O055, MGC11256 | |
CRELD2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9CYA0 | |
79174 | |
Mouse myeloma cell line NS0-derived recombinant mouse CRELD2 Ser23-Leu350 Accession # Q9CYA0 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse CRELD2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CRELD2 is observed and less than 1% cross-reactivity with recombinant mouse CRELD1 is observed. |
For Research Use Only