missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DCAR/CLEC4B Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2757-SP
This item is not returnable.
View return policy
Description
DCAR/CLEC4B Polyclonal specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
DCAR/CLEC4B | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Aplra2, CLEC4B, CLECSF3, LSLCL, P47, SCGF | |
Goat | |
Affinity Purified | |
RUO | |
6320 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q7TS58 | |
Clec4b1 | |
Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B Gln42-Leu209 Accession # Q7TS58 | |
25 μg | |
Primary | |
Detects mouse DCAR/CLEC4B in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse DCIR is observed. | |
Mouse | |
IgG |
For Research Use Only