missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DCIR/CLEC4A Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
£147.94 - £333.68
Specifications
Antigen | DCIR/CLEC4A |
---|---|
Clone | 320511 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788528
|
R&D Systems
MAB2617-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18140241
|
R&D Systems
MAB2617 |
100 μg |
£333.68
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Mouse samples. It is validated for Western Blot.Specifications
DCIR/CLEC4A | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9QZ15 | |
50856 | |
Mouse myeloma cell line NS0-derived recombinant mouse DCIR/CLEC4A isoform 1 Gln70-Leu238 Accession # Q9QZ15 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse DCIR/CLEC4A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) DCIR or recombinant mouse DCAR is observed. |
320511 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD367, CLEC4A, CLECSF6, CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor, LLIR | |
Clec4a2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only