missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DcTRAILR1/TNFRSF23 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2378-SP
This item is not returnable.
View return policy
Description
DcTRAILR1/TNFRSF23 Polyclonal specifically detects DcTRAILR1/TNFRSF23 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DcTRAILR1/TNFRSF23 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Tnfrsf23 | |
Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23 Met30-Ser156 Accession # Q9ER63 | |
25 μg | |
Primary | |
Detects mouse DcTRAIL R1/TNFRSF23 in direct ELISAs and Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) DcTRAIL R2 is observed and less than 1% cross-reactivity with rmTRAIL R2, recombinant human (rh) TRAIL R3, and rhTRAIL R4 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9ER63 | |
Goat | |
Affinity Purified | |
RUO | |
8718 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only